Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
Chromosome Location
Chr.9: 17906574 - 18910950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040272.5 NP_001035362.3 11 - 12 1492 102
NM_052866.4 NP_443098.3 11 - 12 1492 102
XM_011518063.2 XP_011516365.1 13 - 14 1630 102
XM_011518064.2 XP_011516366.1 12 - 13 1745 102
XM_011518067.1 XP_011516369.1 3 - 4 360 102
XM_017015310.1 XP_016870799.1 12 - 13 8267 102
XM_017015311.1 XP_016870800.1 12 - 13 1579 102
XM_017015312.1 XP_016870801.1 11 - 12 1695 102
XM_017015313.1 XP_016870802.1 12 - 13 1393 102
XM_017015314.1 XP_016870803.1 13 - 14 1630 102
GenBank mRNA AF176313.1 - 11 - 12 1390 102
AF251058.1 - 6 - 7 703 102
AK290667.1 - 11 - 12 1478 102
AK291956.1 - 11 - 12 1432 102
AY358327.1 - 11 - 12 1472 102

View Full Product Details