The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040272.5 | NP_001035362.3 | 13 - 14 | 1724 | 110 |
XM_011518063.2 | XP_011516365.1 | 15 - 16 | 1862 | 110 | |
XM_011518064.2 | XP_011516366.1 | 14 - 15 | 1977 | 110 | |
XM_011518067.1 | XP_011516369.1 | 5 - 6 | 592 | 110 | |
XM_011518068.2 | XP_011516370.1 | 3 - 4 | 1195 | 110 | |
XM_011518070.2 | XP_011516372.1 | 3 - 4 | 360 | 110 | |
XM_017015310.1 | XP_016870799.1 | 14 - 15 | 8499 | 110 | |
XM_017015311.1 | XP_016870800.1 | 14 - 15 | 1811 | 110 | |
XM_017015312.1 | XP_016870801.1 | 13 - 14 | 1927 | 110 | |
XM_017015313.1 | XP_016870802.1 | 14 - 15 | 1625 | 110 | |
XM_017015314.1 | XP_016870803.1 | 15 - 16 | 1862 | 110 | |
GenBank mRNA | AF251058.1 | - | 8 - 9 | 935 | 110 |