Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
Chromosome Location
Chr.9: 17906574 - 18910950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040272.5 NP_001035362.3 6 - 7 835 153
NM_052866.4 NP_443098.3 6 - 7 835 153
XM_011518063.2 XP_011516365.1 8 - 9 973 153
XM_011518064.2 XP_011516366.1 7 - 8 1088 153
XM_017015310.1 XP_016870799.1 7 - 8 7610 153
XM_017015311.1 XP_016870800.1 8 - 9 973 153
XM_017015312.1 XP_016870801.1 7 - 8 1089 153
XM_017015313.1 XP_016870802.1 7 - 8 736 153
XM_017015314.1 XP_016870803.1 8 - 9 973 153
GenBank mRNA AF176313.1 - 6 - 7 733 153
AK290667.1 - 6 - 7 821 153
AK291956.1 - 6 - 7 775 153
BC030262.1 - 6 - 7 747 153

View Full Product Details