Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
Chromosome Location
Chr.9: 17906574 - 18910950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040272.5 NP_001035362.3 7 - 8 987 94
NM_052866.4 NP_443098.3 7 - 8 987 94
XM_011518063.2 XP_011516365.1 9 - 10 1125 94
XM_011518064.2 XP_011516366.1 8 - 9 1240 94
XM_017015310.1 XP_016870799.1 8 - 9 7762 94
XM_017015311.1 XP_016870800.1 9 - 10 1125 94
XM_017015312.1 XP_016870801.1 8 - 9 1241 94
XM_017015313.1 XP_016870802.1 8 - 9 888 94
XM_017015314.1 XP_016870803.1 9 - 10 1125 94
GenBank mRNA AF176313.1 - 7 - 8 885 94
AF251058.1 - 2 - 3 198 94
AK290667.1 - 7 - 8 973 94
AK291956.1 - 7 - 8 927 94
AY358327.1 - 7 - 8 967 94
BC030262.1 - 7 - 8 899 94

View Full Product Details