The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003388.4 | NP_003379.3 | 13 - 14 | 3045 | 72 |
NM_032421.2 | NP_115797.1 | 12 - 13 | 2940 | 72 | |
GenBank mRNA | AB006629.2 | - | 12 - 13 | 2940 | 72 |
BC150332.1 | - | 12 - 13 | 2951 | 72 |