The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003388.4 | NP_003379.3 | 2 - 3 | 447 | 57 |
NM_032421.2 | NP_115797.1 | 2 - 3 | 447 | 57 | |
GenBank mRNA | AB006629.2 | - | 2 - 3 | 447 | 57 |
BC150332.1 | - | 2 - 3 | 458 | 57 |