The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321427.1 | NP_001308356.1 | 4 - 5 | 1028 | 81 |
NM_001321428.1 | NP_001308357.1 | 3 - 4 | 439 | 81 | |
NM_001321429.1 | NP_001308358.1 | 4 - 5 | 615 | 81 | |
NM_001321430.1 | NP_001308359.1 | 4 - 5 | 555 | 81 | |
NM_001321432.1 | NP_001308361.1 | 3 - 4 | 379 | 81 | |
NM_001321433.1 | NP_001308362.1 | 4 - 5 | 552 | 81 | |
NM_001321435.1 | NP_001308364.1 | 4 - 5 | 626 | 81 | |
NM_001321436.1 | NP_001308365.1 | 4 - 5 | 630 | 81 | |
NM_022152.5 | NP_071435.2 | 3 - 4 | 454 | 81 | |
NR_135643.1 | 2 - 3 | 212 | 81 | ||
XM_011511627.1 | XP_011509929.1 | 3 - 4 | 807 | 81 | |
GenBank mRNA | AB257295.1 | - | 3 - 4 | 515 | 81 |
AF193045.1 | - | 3 - 4 | 368 | 81 | |
AK027755.1 | - | 1 - 2 | 102 | 81 | |
AK090618.1 | - | 4 - 5 | 1026 | 81 | |
AK093911.1 | - | 1 - 2 | 966 | 81 | |
AK130380.1 | - | 3 - 4 | 412 | 81 | |
AK130713.1 | - | 3 - 4 | 390 | 81 | |
AK298123.1 | - | 4 - 5 | 226 | 81 | |
AK298500.1 | - | 2 - 3 | 148 | 81 | |
BC007980.2 | - | 3 - 4 | 368 | 81 | |
BC013428.1 | - | 3 - 4 | 432 | 81 | |
BC026348.1 | - | 3 - 4 | 389 | 81 | |
BC026693.2 | - | 3 - 4 | 400 | 81 | |
CF994144.1 | - | 3 - 4 | 416 | 81 |