The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145107.1 | NP_001138579.1 | 14 - 15 | 1713 | 140 |
NM_001145108.1 | NP_001138580.1 | 13 - 14 | 2033 | 140 | |
NM_001145109.1 | NP_001138581.1 | 13 - 14 | 1538 | 140 | |
NM_001145110.1 | NP_001138582.1 | 14 - 15 | 1599 | 140 | |
NM_006159.2 | NP_006150.1 | 14 - 15 | 1640 | 140 | |
XM_005268905.3 | XP_005268962.1 | 14 - 15 | 1793 | 140 | |
XM_011538396.1 | XP_011536698.1 | 15 - 16 | 1589 | 140 | |
XM_017019341.1 | XP_016874830.1 | 14 - 15 | 1969 | 140 | |
XM_017019342.1 | XP_016874831.1 | 14 - 15 | 1652 | 140 | |
XM_017019343.1 | XP_016874832.1 | 14 - 15 | 1808 | 140 | |
XM_017019344.1 | XP_016874833.1 | 15 - 16 | 1604 | 140 | |
GenBank mRNA | AK095637.1 | - | 14 - 15 | 2024 | 140 |
AK297521.1 | - | 15 - 16 | 1628 | 140 | |
AK299277.1 | - | 13 - 14 | 1494 | 140 | |
AK299733.1 | - | 14 - 15 | 1713 | 140 | |
AK309825.1 | - | 15 - 16 | 1624 | 140 | |
AK315960.1 | - | 13 - 14 | 1467 | 140 | |
AK316058.1 | - | 14 - 15 | 1960 | 140 | |
BC020544.1 | - | 13 - 14 | 1740 | 140 | |
D83018.1 | - | 14 - 15 | 1534 | 140 | |
D89629.1 | - | 13 - 14 | 1435 | 140 |