The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145107.1 | NP_001138579.1 | 16 - 17 | 1938 | 80 |
NM_001145108.1 | NP_001138580.1 | 15 - 16 | 2258 | 80 | |
NM_001145109.1 | NP_001138581.1 | 15 - 16 | 1763 | 80 | |
NM_001145110.1 | NP_001138582.1 | 16 - 17 | 1824 | 80 | |
NM_006159.2 | NP_006150.1 | 16 - 17 | 1865 | 80 | |
XM_005268905.3 | XP_005268962.1 | 16 - 17 | 2018 | 80 | |
XM_011538396.1 | XP_011536698.1 | 17 - 18 | 1814 | 80 | |
XM_017019341.1 | XP_016874830.1 | 16 - 17 | 2194 | 80 | |
XM_017019342.1 | XP_016874831.1 | 16 - 17 | 1877 | 80 | |
XM_017019343.1 | XP_016874832.1 | 16 - 17 | 2033 | 80 | |
XM_017019344.1 | XP_016874833.1 | 17 - 18 | 1829 | 80 | |
GenBank mRNA | AK095637.1 | - | 16 - 17 | 2249 | 80 |
AK295125.1 | - | 16 - 17 | 1824 | 80 | |
AK297521.1 | - | 17 - 18 | 1853 | 80 | |
AK299277.1 | - | 15 - 16 | 1719 | 80 | |
AK299733.1 | - | 16 - 17 | 1938 | 80 | |
AK315960.1 | - | 15 - 16 | 1692 | 80 | |
AK316058.1 | - | 16 - 17 | 2185 | 80 | |
BC020544.1 | - | 15 - 16 | 1965 | 80 | |
D83018.1 | - | 16 - 17 | 1759 | 80 | |
D89629.1 | - | 15 - 16 | 1660 | 80 |