The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145107.1 | NP_001138579.1 | 7 - 8 | 956 | 67 |
NM_001145108.1 | NP_001138580.1 | 6 - 7 | 1276 | 67 | |
NM_001145109.1 | NP_001138581.1 | 6 - 7 | 781 | 67 | |
NM_001145110.1 | NP_001138582.1 | 7 - 8 | 842 | 67 | |
NM_006159.2 | NP_006150.1 | 7 - 8 | 883 | 67 | |
XM_005268905.3 | XP_005268962.1 | 7 - 8 | 1036 | 67 | |
XM_011538396.1 | XP_011536698.1 | 8 - 9 | 832 | 67 | |
GenBank mRNA | AK056082.1 | - | 6 - 7 | 1127 | 67 |
AK095637.1 | - | 7 - 8 | 1267 | 67 | |
AK295125.1 | - | 7 - 8 | 842 | 67 | |
AK297521.1 | - | 8 - 9 | 871 | 67 | |
AK299733.1 | - | 7 - 8 | 956 | 67 | |
AK309825.1 | - | 8 - 9 | 867 | 67 | |
AK315960.1 | - | 6 - 7 | 710 | 67 | |
AK316058.1 | - | 7 - 8 | 1203 | 67 | |
BC020544.1 | - | 6 - 7 | 983 | 67 | |
D83018.1 | - | 7 - 8 | 777 | 67 | |
D89629.1 | - | 6 - 7 | 678 | 67 |