The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003970.3 | NP_003961.3 | 19 - 20 | 2574 | 91 |
XM_006716237.1 | XP_006716300.1 | 19 - 20 | 2539 | 91 | |
GenBank mRNA | AK297890.1 | - | 6 - 7 | 892 | 91 |
BC052969.1 | - | 19 - 20 | 2527 | 91 | |
X69089.1 | - | 19 - 20 | 2484 | 91 |