Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYOM2
Entrez Gene ID
9172
Gene Name
myomesin 2
Gene Aliases
TTNAP
Chromosome Location
Chr.8: 2045043 - 2145456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003970.3 NP_003961.3 24 - 25 3187 93
XM_006716237.1 XP_006716300.1 24 - 25 3152 93
GenBank mRNA AK297890.1 - 11 - 12 1505 93
BC052969.1 - 24 - 25 3140 93
BX501741.1 - 4 - 5 376 93
DA900083.1 - 3 - 4 177 93
X69089.1 - 24 - 25 3097 93

View Full Product Details