Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYOM2
Entrez Gene ID
9172
Gene Name
myomesin 2
Gene Aliases
TTNAP
Chromosome Location
Chr.8: 2045043 - 2145456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003970.3 NP_003961.3 26 - 27 3468 80
XM_006716237.1 XP_006716300.1 26 - 27 3433 80
GenBank mRNA AK297890.1 - 13 - 14 1786 80
BC052969.1 - 26 - 27 3421 80
BP260135.1 - 1 - 2 141 80
BP260324.1 - 1 - 2 145 80
DA900083.1 - 5 - 6 458 80
X69089.1 - 26 - 27 3378 80

View Full Product Details