The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003970.3 | NP_003961.3 | 31 - 32 | 3837 | 96 |
XM_006716237.1 | XP_006716300.1 | 31 - 32 | 3802 | 96 | |
GenBank mRNA | AK297890.1 | - | 18 - 19 | 2155 | 96 |
BC052969.1 | - | 31 - 32 | 3790 | 96 | |
BP260135.1 | - | 6 - 7 | 510 | 96 | |
BP260324.1 | - | 6 - 7 | 514 | 96 | |
X69089.1 | - | 31 - 32 | 3747 | 96 |