Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOM2
Entrez Gene ID
9172
Gene Name
myomesin 2
Gene Aliases
TTNAP
Chromosome Location
Chr.8: 2045043 - 2145456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003970.3 NP_003961.3 32 - 33 3933 153
XM_006716237.1 XP_006716300.1 32 - 33 3898 153
GenBank mRNA AK297890.1 - 19 - 20 2251 153
BC052969.1 - 32 - 33 3886 153
X69089.1 - 32 - 33 3843 153

View Full Product Details