The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300927.1 | NP_001287856.1 | 4 - 4 | 6546 | 143 |
NM_001308366.1 | NP_001295295.1 | 5 - 5 | 6739 | 143 | |
NM_017712.3 | NP_060182.1 | 5 - 5 | 6779 | 143 | |
XM_006722783.3 | XP_006722846.1 | 5 - 5 | 6679 | 143 | |
XM_011528115.1 | XP_011526417.1 | 4 - 4 | 6704 | 143 | |
XM_017026913.1 | XP_016882402.1 | 4 - 4 | 6629 | 143 | |
GenBank mRNA | AK025948.1 | - | 1 - 1 | 1317 | 143 |
BC004942.1 | - | 1 - 1 | 690 | 143 |