Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL25
Entrez Gene ID
84190
Gene Name
methyltransferase like 25
Gene Aliases
C12orf26
Chromosome Location
Chr.12: 82358497 - 82479239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319675.1 NP_001306604.1 9 - 10 1733 99
NM_032230.2 NP_115606.2 9 - 10 1641 99
XM_011538827.2 XP_011537129.1 8 - 9 1429 99
XM_017020020.1 XP_016875509.1 10 - 11 1704 99
XM_017020021.1 XP_016875510.1 9 - 10 2624 99
GenBank mRNA BC029120.1 - 9 - 10 1593 99

View Full Product Details