Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGD6
Entrez Gene ID
55785
Gene Name
FYVE, RhoGEF and PH domain containing 6
Gene Aliases
ZFYVE24
Chromosome Location
Chr.12: 95076749 - 95217464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018351.3 NP_060821.3 15 - 16 3817 114
GenBank mRNA AB037783.1 - 14 - 15 1590 114
AK002045.1 - 8 - 9 608 114
AK124904.1 - 15 - 16 3817 114
AK128492.1 - 14 - 15 2694 114
BC013319.1 - 9 - 10 700 114
BC125225.1 - 14 - 15 1220 114
BC125226.1 - 14 - 15 1220 114

View Full Product Details