The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002945.3 | NP_002936.1 | 16 - 17 | 1859 | 92 |
GenBank mRNA | AB209732.1 | - | 16 - 17 | 1787 | 92 |
AK289704.1 | - | 16 - 17 | 1825 | 92 | |
BC018126.1 | - | 16 - 17 | 1787 | 92 | |
M63488.1 | - | 16 - 17 | 1813 | 92 |