Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGBL1
Entrez Gene ID
9358
Gene Name
integrin subunit beta like 1
Gene Aliases
OSCP, TIED
Chromosome Location
Chr.13: 101452593 - 101720857 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271754.1 NP_001258683.1 7 - 8 1162 66
NM_001271755.1 NP_001258684.1 7 - 8 1233 66
NM_001271756.1 NP_001258685.1 7 - 8 872 66
NM_004791.2 NP_004782.1 8 - 9 1380 66
XM_005254100.4 XP_005254157.1 8 - 9 1338 66
GenBank mRNA AB008375.1 - 9 - 10 1244 66
AF072752.1 - 8 - 9 1358 66
AK075136.1 - 2 - 3 281 66
AK095875.1 - 8 - 9 1020 66
AK291340.1 - 8 - 9 1300 66
AK297744.1 - 7 - 8 1088 66
AK298571.1 - 7 - 8 872 66
AK308194.1 - 7 - 8 1150 66
BC036788.1 - 8 - 9 1301 66

View Full Product Details