The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160706.1 | NP_001154178.1 | 12 - 13 | 898 | 179 |
NM_003843.3 | NP_003834.3 | 12 - 13 | 904 | 119 | |
NM_144777.2 | NP_659001.2 | 13 - 14 | 964 | 179 | |
XM_005266578.1 | XP_005266635.1 | 12 - 13 | 875 | 179 | |
XM_006719884.1 | XP_006719947.1 | 12 - 13 | 844 | 179 | |
XM_011535281.1 | XP_011533583.1 | 13 - 14 | 910 | 179 | |
XM_011535282.1 | XP_011533584.1 | 13 - 14 | 910 | 179 | |
XM_011535283.1 | XP_011533585.1 | 13 - 14 | 910 | 179 | |
XM_011535284.1 | XP_011533586.1 | 13 - 14 | 910 | 179 | |
XM_011535285.1 | XP_011533587.1 | 12 - 13 | 920 | 179 | |
XM_011535286.1 | XP_011533588.1 | 13 - 14 | 910 | 179 | |
XM_011535287.1 | XP_011533589.1 | 13 - 14 | 910 | 179 | |
XM_011535288.1 | XP_011533590.1 | 11 - 12 | 774 | 179 | |
XM_011535290.1 | XP_011533592.1 | 13 - 14 | 910 | 179 | |
XM_011535291.1 | XP_011533593.1 | 13 - 14 | 910 | 179 | |
XM_017020805.1 | XP_016876294.1 | 13 - 14 | 910 | 179 | |
GenBank mRNA | AF045941.1 | - | 12 - 13 | 820 | 119 |
AK025320.1 | - | 13 - 14 | 878 | 179 | |
AK222720.1 | - | 13 - 14 | 878 | 179 | |
AK301659.1 | - | 12 - 13 | 812 | 179 | |
BC020726.1 | - | 11 - 12 | 785 | 119 | |
BC047536.1 | - | 12 - 13 | 979 | 119 |