Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCEL
Entrez Gene ID
8796
Gene Name
sciellin
Gene Aliases
-
Chromosome Location
Chr.13: 77535674 - 77645263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160706.1 NP_001154178.1 3 - 4 327 139
NM_003843.3 NP_003834.3 3 - 4 327 139
NM_144777.2 NP_659001.2 3 - 4 327 139
XM_005266578.1 XP_005266635.1 3 - 4 307 139
XM_006719882.1 XP_006719945.1 3 - 4 273 139
XM_006719884.1 XP_006719947.1 3 - 4 273 139
XM_006719885.1 XP_006719948.1 3 - 4 273 139
XM_011535281.1 XP_011533583.1 3 - 4 273 139
XM_011535282.1 XP_011533584.1 3 - 4 273 139
XM_011535283.1 XP_011533585.1 3 - 4 273 139
XM_011535284.1 XP_011533586.1 3 - 4 273 139
XM_011535285.1 XP_011533587.1 3 - 4 322 139
XM_011535286.1 XP_011533588.1 3 - 4 273 139
XM_011535287.1 XP_011533589.1 3 - 4 273 139
XM_011535288.1 XP_011533590.1 3 - 4 245 139
XM_011535289.1 XP_011533591.1 3 - 4 273 139
XM_011535290.1 XP_011533592.1 3 - 4 273 139
XM_011535291.1 XP_011533593.1 3 - 4 273 139
XM_017020805.1 XP_016876294.1 3 - 4 273 139
GenBank mRNA AF045941.1 - 3 - 4 243 139
AK025320.1 - 3 - 4 241 139
AK222720.1 - 3 - 4 241 139
AK301659.1 - 3 - 4 241 139
BC020726.1 - 3 - 4 277 139
BC047536.1 - 3 - 4 402 139

View Full Product Details