The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025099.5 | NP_079375.3 | 13 - 14 | 2440 | 65 |
NR_046431.1 | 13 - 14 | 2335 | 65 | ||
XM_006721577.3 | XP_006721640.1 | 12 - 13 | 2295 | 65 | |
XM_006721578.3 | XP_006721641.1 | 13 - 14 | 2424 | 65 | |
XM_011524010.2 | XP_011522312.1 | 13 - 14 | 2319 | 65 | |
XM_011524011.2 | XP_011522313.1 | 12 - 13 | 2189 | 65 | |
GenBank mRNA | AK025823.1 | - | 3 - 4 | 583 | 65 |
AK091077.1 | - | 13 - 14 | 2391 | 65 | |
AK225375.1 | - | 3 - 4 | 583 | 65 | |
AK310636.1 | - | 2 - 3 | 609 | 65 | |
AL831955.1 | - | 13 - 14 | 2282 | 65 | |
BC026057.2 | - | 4 - 5 | 581 | 65 | |
BC110373.1 | - | 5 - 6 | 807 | 65 | |
BC111783.1 | - | 13 - 14 | 2391 | 65 | |
BX406692.2 | - | 1 - 2 | 193 | 65 | |
DQ451688.1 | - | 13 - 14 | 2381 | 65 | |
FJ790670.1 | - | 13 - 14 | 2381 | 65 |