The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025099.5 | NP_079375.3 | 14 - 15 | 2533 | 112 |
NR_046431.1 | 14 - 15 | 2428 | 112 | ||
XM_006721577.3 | XP_006721640.1 | 13 - 14 | 2388 | 112 | |
XM_006721578.3 | XP_006721641.1 | 14 - 15 | 2517 | 112 | |
XM_011524010.2 | XP_011522312.1 | 14 - 15 | 2412 | 112 | |
XM_011524011.2 | XP_011522313.1 | 13 - 14 | 2282 | 112 | |
GenBank mRNA | AK025823.1 | - | 4 - 5 | 676 | 112 |
AK091077.1 | - | 14 - 15 | 2484 | 112 | |
AK225375.1 | - | 4 - 5 | 676 | 112 | |
AK310636.1 | - | 3 - 4 | 702 | 112 | |
AL831955.1 | - | 14 - 15 | 2375 | 112 | |
BC026057.2 | - | 5 - 6 | 674 | 112 | |
BC110373.1 | - | 6 - 7 | 900 | 112 | |
BC111783.1 | - | 14 - 15 | 2484 | 112 | |
DQ451688.1 | - | 14 - 15 | 2474 | 112 |