The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031848.1 | NP_001027018.1 | 4 - 5 | 572 | 98 |
NM_001276490.1 | NP_001263419.1 | 3 - 4 | 394 | 98 | |
NM_002640.3 | NP_002631.3 | 4 - 5 | 572 | 98 | |
NM_198833.1 | NP_942130.1 | 4 - 5 | 589 | 98 | |
XM_006722484.2 | XP_006722547.1 | 4 - 5 | 520 | 98 | |
XM_011526025.1 | XP_011524327.1 | 4 - 5 | 573 | 98 | |
XM_017025790.1 | XP_016881279.1 | 4 - 5 | 573 | 98 | |
XM_017025791.1 | XP_016881280.1 | 4 - 5 | 517 | 98 | |
GenBank mRNA | AK313782.1 | - | 4 - 5 | 489 | 98 |
AL708543.1 | - | 4 - 5 | 537 | 98 | |
BC034528.1 | - | 4 - 5 | 482 | 98 | |
BQ435307.1 | - | 4 - 5 | 595 | 98 | |
BX571754.1 | - | 4 - 5 | 480 | 98 | |
DC384755.1 | - | 3 - 4 | 328 | 98 | |
L40377.1 | - | 4 - 5 | 503 | 98 | |
U92984.1 | - | 4 - 5 | 481 | 98 |