The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166136.1 | NP_001159608.1 | 18 - 19 | 3761 | 70 |
NM_147127.4 | NP_667338.3 | 18 - 19 | 3323 | 70 | |
XM_017007735.1 | XP_016863224.1 | 18 - 19 | 3334 | 70 | |
XM_017007736.1 | XP_016863225.1 | 19 - 20 | 3312 | 70 | |
XM_017007737.1 | XP_016863226.1 | 19 - 20 | 3199 | 70 | |
XM_017007739.1 | XP_016863228.1 | 19 - 20 | 3357 | 70 | |
GenBank mRNA | AB083067.1 | - | 17 - 18 | 3029 | 70 |
AY152402.1 | - | 18 - 19 | 3758 | 70 | |
AY152403.1 | - | 18 - 19 | 3758 | 70 | |
AY185210.1 | - | 18 - 19 | 3323 | 70 | |
BC143836.1 | - | 19 - 20 | 3362 | 70 |