Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EVC2
Entrez Gene ID
132884
Gene Name
EvC ciliary complex subunit 2
Gene Aliases
LBN, WAD
Chromosome Location
Chr.4: 5531921 - 5709548 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166136.1 NP_001159608.1 20 - 21 4051 119
NM_147127.4 NP_667338.3 20 - 21 3613 119
XM_017007735.1 XP_016863224.1 20 - 21 3624 119
XM_017007736.1 XP_016863225.1 21 - 22 3602 119
XM_017007737.1 XP_016863226.1 21 - 22 3489 119
XM_017007739.1 XP_016863228.1 21 - 22 3647 119
GenBank mRNA AB083067.1 - 19 - 20 3319 119
AY152402.1 - 20 - 21 4048 119
AY152403.1 - 20 - 21 4048 119
AY185210.1 - 20 - 21 3613 119
BC143836.1 - 21 - 22 3652 119

View Full Product Details