The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166136.1 | NP_001159608.1 | 21 - 22 | 4144 | 97 |
NM_147127.4 | NP_667338.3 | 21 - 22 | 3706 | 97 | |
XM_017007736.1 | XP_016863225.1 | 22 - 23 | 3695 | 97 | |
XM_017007737.1 | XP_016863226.1 | 22 - 23 | 3582 | 97 | |
XM_017007739.1 | XP_016863228.1 | 22 - 23 | 3740 | 97 | |
GenBank mRNA | AB083067.1 | - | 20 - 21 | 3412 | 97 |
AY152402.1 | - | 21 - 22 | 4141 | 97 | |
AY185210.1 | - | 21 - 22 | 3706 | 97 | |
BC143836.1 | - | 22 - 23 | 3745 | 97 |