The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166136.1 | NP_001159608.1 | 5 - 6 | 1197 | 79 |
NM_147127.4 | NP_667338.3 | 5 - 6 | 759 | 79 | |
XM_017007735.1 | XP_016863224.1 | 5 - 6 | 770 | 79 | |
XM_017007736.1 | XP_016863225.1 | 6 - 7 | 748 | 79 | |
XM_017007737.1 | XP_016863226.1 | 6 - 7 | 635 | 79 | |
XM_017007738.1 | XP_016863227.1 | 5 - 6 | 770 | 79 | |
XM_017007739.1 | XP_016863228.1 | 6 - 7 | 801 | 79 | |
GenBank mRNA | AB083067.1 | - | 4 - 5 | 465 | 79 |
AY152402.1 | - | 5 - 6 | 1194 | 79 | |
AY152403.1 | - | 5 - 6 | 1194 | 79 | |
AY185210.1 | - | 5 - 6 | 759 | 79 | |
BC143836.1 | - | 6 - 7 | 806 | 79 |