The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015315.4 | NP_056130.2 | 18 - 19 | 2868 | 76 |
XM_005268404.3 | XP_005268461.1 | 18 - 19 | 3258 | 76 | |
XM_005268406.3 | XP_005268463.1 | 18 - 19 | 3159 | 76 | |
XM_005268407.3 | XP_005268464.1 | 18 - 19 | 3014 | 76 | |
XM_005268408.3 | XP_005268465.1 | 18 - 19 | 2751 | 76 | |
XM_006714775.2 | XP_006714838.1 | 18 - 19 | 2667 | 76 | |
XM_006714776.3 | XP_006714839.1 | 18 - 19 | 2772 | 76 | |
XM_011537614.1 | XP_011535916.1 | 18 - 19 | 3180 | 76 | |
XM_011537615.1 | XP_011535917.1 | 17 - 18 | 3057 | 76 | |
XM_011537616.1 | XP_011535918.1 | 18 - 19 | 2766 | 76 | |
XM_011537617.1 | XP_011535919.1 | 18 - 19 | 2685 | 76 | |
XM_011537618.2 | XP_011535920.1 | 18 - 19 | 2819 | 76 | |
XM_017009283.1 | XP_016864772.1 | 18 - 19 | 2915 | 76 | |
XM_017009284.1 | XP_016864773.1 | 17 - 18 | 2813 | 76 | |
GenBank mRNA | AB018274.1 | - | 19 - 20 | 3075 | 76 |
AL133034.1 | - | 9 - 10 | 1465 | 76 | |
BC001460.2 | - | 18 - 19 | 2868 | 76 | |
BC033856.1 | - | 6 - 7 | 746 | 76 |