The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300860.1 | NP_001287789.1 | 12 - 13 | 1745 | 141 |
NM_014829.3 | NP_055644.2 | 12 - 13 | 1745 | 141 | |
GenBank mRNA | AB018344.1 | - | 12 - 13 | 1684 | 141 |
AF106680.1 | - | 12 - 13 | 1685 | 141 | |
AK291788.1 | - | 12 - 13 | 1698 | 141 | |
BC012304.2 | - | 12 - 13 | 1675 | 141 |