Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STX8
Entrez Gene ID
9482
Gene Name
syntaxin 8
Gene Aliases
CARB
Chromosome Location
Chr.17: 9250471 - 9575958 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004853.2 NP_004844.1 2 - 3 262 138
GenBank mRNA AF036715.1 - 2 - 3 112 138
AF062077.1 - 2 - 3 262 138
AF115323.1 - 2 - 3 262 138
BC009713.1 - 2 - 3 135 138
BT007319.1 - 2 - 3 112 138

View Full Product Details