The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042.2 | NP_001033.1 | 4 - 5 | 652 | 90 |
GenBank mRNA | AL521328.3 | - | 4 - 5 | 651 | 90 |
BC034387.1 | - | 4 - 5 | 652 | 90 | |
BC069615.1 | - | 4 - 5 | 547 | 90 | |
BC069621.1 | - | 4 - 5 | 547 | 90 | |
BC113592.1 | - | 4 - 5 | 479 | 90 | |
BC126164.1 | - | 4 - 5 | 479 | 90 | |
M20747.1 | - | 4 - 5 | 597 | 90 |