The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042.2 | NP_001033.1 | 5 - 6 | 764 | 116 |
GenBank mRNA | AK300899.1 | - | 5 - 6 | 831 | 116 |
AK313372.1 | - | 5 - 6 | 778 | 116 | |
AL521328.3 | - | 5 - 6 | 763 | 116 | |
BC034387.1 | - | 5 - 6 | 764 | 116 | |
BC069615.1 | - | 5 - 6 | 659 | 116 | |
BC069621.1 | - | 5 - 6 | 659 | 116 | |
BC113592.1 | - | 5 - 6 | 591 | 116 | |
BC126164.1 | - | 5 - 6 | 591 | 116 | |
M20747.1 | - | 5 - 6 | 709 | 116 |