The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042.2 | NP_001033.1 | 6 - 7 | 927 | 73 |
GenBank mRNA | AK300899.1 | - | 6 - 7 | 994 | 73 |
AK313372.1 | - | 6 - 7 | 941 | 73 | |
BC034387.1 | - | 6 - 7 | 927 | 73 | |
BC069615.1 | - | 6 - 7 | 822 | 73 | |
BC069621.1 | - | 6 - 7 | 822 | 73 | |
BC113592.1 | - | 6 - 7 | 754 | 73 | |
BC126164.1 | - | 6 - 7 | 754 | 73 | |
M20747.1 | - | 6 - 7 | 872 | 73 |