The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289036.1 | NP_001275965.1 | 8 - 9 | 1390 | 155 |
NM_001318522.1 | NP_001305451.1 | 6 - 7 | 1106 | 155 | |
NM_014410.4 | NP_055225.1 | 6 - 7 | 1145 | 155 | |
NM_199167.1 | NP_954636.1 | 6 - 7 | 1071 | 155 | |
XM_005258103.3 | XP_005258160.1 | 8 - 9 | 1295 | 155 | |
XM_006722322.2 | XP_006722385.1 | 7 - 8 | 1263 | 155 | |
XM_011525648.2 | XP_011523950.1 | 8 - 9 | 1338 | 155 | |
XM_011525649.1 | XP_011523951.1 | 8 - 9 | 1338 | 155 | |
XM_011525651.2 | XP_011523953.1 | 7 - 8 | 2060 | 155 | |
XM_011525652.1 | XP_011523954.1 | 8 - 9 | 1414 | 155 | |
XM_011525653.2 | XP_011523955.1 | 6 - 7 | 1035 | 155 | |
XM_017025707.1 | XP_016881196.1 | 9 - 10 | 2800 | 155 | |
GenBank mRNA | AF395889.1 | - | 6 - 7 | 1060 | 155 |
AK299319.1 | - | 8 - 9 | 1365 | 155 | |
BC025381.2 | - | 6 - 7 | 1145 | 155 | |
D63813.1 | - | 6 - 7 | 1051 | 155 |