Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLAMF6
Entrez Gene ID
114836
Gene Name
SLAM family member 6
Gene Aliases
CD352, KALI, KALIb, Ly108, NTB-A, NTBA, SF2000
Chromosome Location
Chr.1: 160485030 - 160523268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184714.1 NP_001171643.1 3 - 4 711 133
NM_001184715.1 NP_001171644.1 3 - 4 564 133
NM_001184716.1 NP_001171645.1 2 - 3 378 133
NM_052931.4 NP_443163.1 3 - 4 711 133
XM_017000215.1 XP_016855704.1 3 - 4 717 133
XM_017000216.1 XP_016855705.1 3 - 4 537 133
XM_017000217.1 XP_016855706.1 2 - 3 379 133
GenBank mRNA AJ277141.1 - 3 - 4 653 133
AJ306388.1 - 3 - 4 650 133
AK125624.1 - 3 - 4 711 133
AK300013.1 - 2 - 3 361 133
AK301026.1 - 2 - 3 368 133
AK303990.1 - 3 - 4 557 133
AK313549.1 - 3 - 4 663 133
AL832854.1 - 3 - 4 692 133
AY358159.1 - 3 - 4 702 133
BC113893.1 - 3 - 4 668 133
BC114495.1 - 3 - 4 668 133

View Full Product Details