Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLAMF6
Entrez Gene ID
114836
Gene Name
SLAM family member 6
Gene Aliases
CD352, KALI, KALIb, Ly108, NTB-A, NTBA, SF2000
Chromosome Location
Chr.1: 160485030 - 160523268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184714.1 NP_001171643.1 6 - 7 956 159
NM_001184715.1 NP_001171644.1 6 - 7 806 159
NM_001184716.1 NP_001171645.1 5 - 6 623 159
NM_052931.4 NP_443163.1 6 - 7 953 159
XM_017000216.1 XP_016855705.1 6 - 7 782 159
XM_017000217.1 XP_016855706.1 5 - 6 621 159
GenBank mRNA AJ277141.1 - 6 - 7 895 159
AJ306388.1 - 6 - 7 895 159
AK125624.1 - 6 - 7 953 159
AK301026.1 - 5 - 6 613 159
AK303990.1 - 6 - 7 799 159
AK313549.1 - 6 - 7 905 159
AL832854.1 - 6 - 7 937 159
AY358159.1 - 6 - 7 947 159
BC113893.1 - 6 - 7 910 159
BC114495.1 - 6 - 7 913 159

View Full Product Details