Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITIH1
Entrez Gene ID
3697
Gene Name
inter-alpha-trypsin inhibitor heavy chain 1
Gene Aliases
H1P, IATIH, IGHEP1, ITI-HC1, ITIH, SHAP
Chromosome Location
Chr.3: 52777586 - 52792068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166434.2 NP_001159906.1 9 - 10 1278 98
NM_001166435.2 NP_001159907.1 7 - 8 1009 98
NM_001166436.2 NP_001159908.1 7 - 8 895 98
NM_002215.3 NP_002206.2 11 - 12 1439 98
GenBank mRNA AF116713.2 - 2 - 3 301 98
AK292750.1 - 11 - 12 1409 98
AK298398.1 - 11 - 12 1438 98
AK298423.1 - 11 - 12 1431 98
AK298455.1 - 9 - 10 1264 98
AK303110.1 - 7 - 8 895 98
AK303156.1 - 7 - 8 1009 98
AK314198.1 - 11 - 12 1409 98
BC069464.1 - 11 - 12 1433 98
M18192.1 - 2 - 3 216 98
X16260.1 - 9 - 10 1188 98
X63652.1 - 11 - 12 1433 98

View Full Product Details