Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITIH1
Entrez Gene ID
3697
Gene Name
inter-alpha-trypsin inhibitor heavy chain 1
Gene Aliases
H1P, IATIH, IGHEP1, ITI-HC1, ITIH, SHAP
Chromosome Location
Chr.3: 52777586 - 52792068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166434.2 NP_001159906.1 15 - 16 1879 57
NM_001166435.2 NP_001159907.1 13 - 14 1610 57
NM_001166436.2 NP_001159908.1 13 - 14 1496 57
NM_002215.3 NP_002206.2 17 - 18 2040 57
GenBank mRNA AF116713.2 - 8 - 9 902 57
AK292750.1 - 17 - 18 2010 57
AK298455.1 - 15 - 16 1865 57
AK303110.1 - 13 - 14 1496 57
AK303156.1 - 13 - 14 1610 57
AK314198.1 - 17 - 18 2010 57
BC069464.1 - 17 - 18 2034 57
M18192.1 - 8 - 9 817 57
X16260.1 - 15 - 16 1789 57
X63652.1 - 17 - 18 2034 57

View Full Product Details