Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LY6G6F
Entrez Gene ID
259215
Gene Name
lymphocyte antigen 6 complex, locus G6F
Gene Aliases
C6orf21, G6f, LY6G6D, NG32
Chromosome Location
Chr.6: 31706907 - 31710595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003693.1 NP_001003693.1 4 - 5 803 91
GenBank mRNA AJ496460.1 - 4 - 5 803 91
BC137212.1 - 4 - 5 847 91
BC137213.1 - 4 - 5 847 91

View Full Product Details