The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000544.3 | NP_000535.3 | 4 - 5 | 862 | 83 |
NM_001290043.1 | NP_001276972.1 | 4 - 5 | 915 | 83 | |
NM_018833.2 | NP_061313.2 | 4 - 5 | 862 | 83 | |
GenBank mRNA | AB073779.1 | - | 3 - 4 | 740 | 83 |
AB208953.1 | - | 4 - 5 | 809 | 83 | |
AF078671.1 | - | 4 - 5 | 819 | 83 | |
AF105151.1 | - | 4 - 5 | 819 | 83 | |
AK222823.1 | - | 3 - 4 | 771 | 83 | |
AK223300.1 | - | 4 - 5 | 822 | 83 | |
AK225657.1 | - | 4 - 5 | 843 | 83 | |
AK225693.1 | - | 4 - 5 | 822 | 83 | |
AK292963.1 | - | 4 - 5 | 845 | 83 | |
BC002751.2 | - | 3 - 4 | 748 | 83 | |
BT009906.1 | - | 3 - 4 | 740 | 83 | |
FM246456.1 | - | 3 - 4 | 740 | 83 | |
M74447.1 | - | 4 - 5 | 836 | 83 | |
M84748.1 | - | 4 - 5 | 768 | 83 | |
U07844.1 | - | 3 - 4 | 550 | 83 | |
Z22935.1 | - | 4 - 5 | 768 | 83 | |
Z22936.1 | - | 4 - 5 | 816 | 83 |