The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000544.3 | NP_000535.3 | 9 - 10 | 1756 | 88 |
NM_001290043.1 | NP_001276972.1 | 9 - 10 | 1809 | 88 | |
NM_018833.2 | NP_061313.2 | 9 - 10 | 1756 | 88 | |
GenBank mRNA | AB073779.1 | - | 8 - 9 | 1634 | 88 |
AB208953.1 | - | 9 - 10 | 1703 | 88 | |
AF078671.1 | - | 9 - 10 | 1713 | 88 | |
AF105151.1 | - | 9 - 10 | 1713 | 88 | |
AF176984.1 | - | 4 - 5 | 689 | 88 | |
AK222823.1 | - | 8 - 9 | 1665 | 88 | |
AK223300.1 | - | 9 - 10 | 1716 | 88 | |
AK225693.1 | - | 9 - 10 | 1716 | 88 | |
AK292963.1 | - | 9 - 10 | 1739 | 88 | |
BC002751.2 | - | 8 - 9 | 1642 | 88 | |
BT009906.1 | - | 8 - 9 | 1634 | 88 | |
FM246456.1 | - | 8 - 9 | 1634 | 88 | |
L09191.1 | - | 1 - 2 | 86 | 88 | |
L09271.1 | - | 1 - 2 | 86 | 88 | |
L10287.1 | - | 1 - 2 | 86 | 88 | |
M74447.1 | - | 9 - 10 | 1730 | 88 | |
M84748.1 | - | 9 - 10 | 1662 | 88 | |
U07844.1 | - | 8 - 9 | 1444 | 88 | |
Z22935.1 | - | 9 - 10 | 1662 | 88 | |
Z22936.1 | - | 9 - 10 | 1710 | 88 |