The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201338.1 | NP_001188267.1 | 3 - 4 | 505 | 74 |
NM_001201339.1 | NP_001188268.1 | 3 - 4 | 505 | 74 | |
NM_001320572.1 | NP_001307501.1 | 3 - 4 | 505 | 74 | |
NM_002967.3 | NP_002958.2 | 3 - 4 | 505 | 74 | |
XM_017027116.1 | XP_016882605.1 | 3 - 4 | 406 | 74 | |
XM_017027117.1 | XP_016882606.1 | 3 - 4 | 406 | 74 | |
XM_017027118.1 | XP_016882607.1 | 3 - 4 | 406 | 74 | |
GenBank mRNA | AB208780.1 | - | 3 - 4 | 422 | 74 |
AK290444.1 | - | 3 - 4 | 401 | 74 | |
AK294718.1 | - | 3 - 4 | 457 | 74 | |
BC020309.1 | - | 3 - 4 | 387 | 74 | |
BC032055.1 | - | 3 - 4 | 384 | 74 | |
BC126219.1 | - | 3 - 4 | 376 | 74 | |
BC143937.1 | - | 3 - 4 | 373 | 74 | |
BC143938.1 | - | 3 - 4 | 376 | 74 | |
BC143939.1 | - | 3 - 4 | 376 | 74 | |
DC349269.1 | - | 3 - 4 | 491 | 74 | |
L43631.1 | - | 3 - 4 | 182 | 74 | |
U72355.1 | - | 3 - 4 | 387 | 74 |