Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF10
Entrez Gene ID
9639
Gene Name
Rho guanine nucleotide exchange factor 10
Gene Aliases
GEF10, SNCV
Chromosome Location
Chr.8: 1823976 - 1958641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308152.1 NP_001295081.1 3 - 4 381 99
NM_001308153.1 NP_001295082.1 4 - 5 470 99
NM_014629.3 NP_055444.2 3 - 4 381 99
XM_005266041.3 XP_005266098.1 3 - 4 320 99
XM_011534767.2 XP_011533069.1 2 - 3 243 99
XM_011534768.1 XP_011533070.1 2 - 3 243 99
XM_011534770.2 XP_011533072.1 2 - 3 243 99
XM_017014003.1 XP_016869492.1 3 - 4 283 99
GenBank mRNA AB002292.2 - 3 - 4 380 99
BC112926.1 - 3 - 4 369 99
CR749570.1 - 4 - 5 431 99

View Full Product Details