The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009566.2 | NP_001009566.1 | 10 - 11 | 2313 | 94 |
NM_014944.4 | NP_055759.3 | 9 - 10 | 2283 | 94 | |
GenBank mRNA | AB020718.1 | - | 10 - 11 | 2313 | 94 |
AF438482.1 | - | 9 - 10 | 2249 | 94 | |
AK289798.1 | - | 9 - 10 | 1491 | 94 | |
AY753301.1 | - | 9 - 10 | 1490 | 94 | |
BC033902.2 | - | 9 - 10 | 1741 | 94 |