Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLOT1
Entrez Gene ID
10211
Gene Name
flotillin 1
Gene Aliases
-
Chromosome Location
Chr.6: 30727709 - 30742851 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318875.1 NP_001305804.1 5 - 6 672 66
NM_005803.3 NP_005794.1 6 - 7 816 66
XM_005248780.3 XP_005248837.1 6 - 7 549 66
XM_006714947.3 XP_006715010.1 5 - 6 653 66
XM_017010157.1 XP_016865646.1 6 - 7 470 66
XM_017010158.1 XP_016865647.1 4 - 5 371 66
GenBank mRNA AF085357.1 - 6 - 7 605 66
AF089750.2 - 6 - 7 641 66
AF117234.1 - 6 - 7 642 66
AK000816.1 - 6 - 7 652 66
AK222530.1 - 6 - 7 652 66
AK301291.1 - 5 - 6 506 66
BC001146.1 - 5 - 6 499 66
BT007077.1 - 5 - 6 477 66
CR456946.1 - 5 - 6 477 66

View Full Product Details