Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTN
Entrez Gene ID
7273
Gene Name
titin
Gene Aliases
CMD1G, CMH9, CMPD4, EOMFC, HMERF, LGMD2J, MYLK5, TMD
Chromosome Location
Chr.2: 178525989 - 178807423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256850.1 NP_001243779.1 14 - 15 2597 108
NM_001267550.2 NP_001254479.2 14 - 15 2597 108
NM_003319.4 NP_003310.4 13 - 14 2459 108
NM_133378.4 NP_596869.4 14 - 15 2597 108
NM_133379.4 NP_596870.2 14 - 15 2597 108
NM_133432.3 NP_597676.3 13 - 14 2459 108
NM_133437.4 NP_597681.4 13 - 14 2459 108
XM_017004819.1 XP_016860308.1 14 - 15 2600 108
XM_017004820.1 XP_016860309.1 14 - 15 2600 108
XM_017004821.1 XP_016860310.1 14 - 15 2597 108
XM_017004822.1 XP_016860311.1 14 - 15 2600 108
XM_017004823.1 XP_016860312.1 14 - 15 2600 108
GenBank mRNA X90568.1 - 13 - 14 2366 108

View Full Product Details