Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTN
Entrez Gene ID
7273
Gene Name
titin
Gene Aliases
CMD1G, CMH9, CMPD4, EOMFC, HMERF, LGMD2J, MYLK5, TMD
Chromosome Location
Chr.2: 178525989 - 178807423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256850.1 NP_001243779.1 36 - 37 8862 83
NM_001267550.2 NP_001254479.2 36 - 37 8862 83
NM_003319.4 NP_003310.4 35 - 36 8724 83
NM_133378.4 NP_596869.4 36 - 37 8862 83
NM_133379.4 NP_596870.2 36 - 37 8862 83
NM_133432.3 NP_597676.3 35 - 36 8724 83
NM_133437.4 NP_597681.4 35 - 36 8724 83
XM_017004819.1 XP_016860308.1 36 - 37 8865 83
XM_017004820.1 XP_016860309.1 36 - 37 8865 83
XM_017004821.1 XP_016860310.1 36 - 37 8862 83
XM_017004822.1 XP_016860311.1 36 - 37 8865 83
XM_017004823.1 XP_016860312.1 36 - 37 8865 83
GenBank mRNA AK129531.1 - 4 - 5 836 83
X90568.1 - 35 - 36 8631 83

View Full Product Details