The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000275.2 | NP_000266.2 | 5 - 6 | 685 | 69 |
NM_001300984.1 | NP_001287913.1 | 5 - 6 | 685 | 69 | |
XM_011521640.2 | XP_011519942.1 | 5 - 6 | 715 | 69 | |
XM_017022255.1 | XP_016877744.1 | 5 - 6 | 666 | 69 | |
XM_017022256.1 | XP_016877745.1 | 5 - 6 | 1163 | 69 | |
XM_017022257.1 | XP_016877746.1 | 5 - 6 | 688 | 69 | |
XM_017022258.1 | XP_016877747.1 | 5 - 6 | 688 | 69 | |
XM_017022259.1 | XP_016877748.1 | 5 - 6 | 688 | 69 | |
XM_017022260.1 | XP_016877749.1 | 5 - 6 | 688 | 69 | |
XM_017022261.1 | XP_016877750.1 | 4 - 5 | 1182 | 69 | |
XM_017022262.1 | XP_016877751.1 | 5 - 6 | 688 | 69 | |
XM_017022263.1 | XP_016877752.1 | 5 - 6 | 688 | 69 | |
XM_017022264.1 | XP_016877753.1 | 5 - 6 | 688 | 69 | |
XM_017022265.1 | XP_016877754.1 | 5 - 6 | 688 | 69 | |
GenBank mRNA | BC012097.1 | - | 5 - 6 | 685 | 69 |
M99564.1 | - | 5 - 6 | 627 | 69 |